Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APRIL/TNFSF13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8843
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8843 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8843 Supplier R&D Systems Supplier No. MAB8843
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APRIL/TNFSF13 Monoclonal specifically detects APRIL/TNFSF13 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen APRIL/TNFSF13
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 670840
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75888
Gene Alias A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TNFSF13, TRDL1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2
Gene Symbols TNFSF13
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293EBNA-derived recombinant human APRIL/TNFSF13 Ala105-Leu250 Accession # O75888
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8741
Test Specificity Detects human APRIL/TNFSF13 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse APRIL, recombinant human (rh) BAFF, rhEDA-A2, rhEDA, rhFas Ligand, rhLIGHT, rhLT a1/b2, rhLT a2/b1, rhOX40 Ligand, rhTNF alpha, rhTRAIL, rhTRANCE, rh4-1BB Ligand, rhVEGI, or rhGITR Ligand is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.