Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APRIL/TNFSF13 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $585.00
Specifications
Antigen | APRIL/TNFSF13 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Classification | Polyclonal |
Description
APRIL/TNFSF13 Polyclonal specifically detects APRIL/TNFSF13 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
APRIL/TNFSF13 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
8741 | |
E. coli-derived recombinant human APRIL/TNFSF13 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human APRIL/TNFSF13 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) Fas Ligand, rhTNF-alpha , rhTRAIL, and rhTRANCE is observed. |
LYOPH | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TNFSF13, TRDL1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
TNFSF13 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title