Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APRIL/TNFSF13 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF884
This item is not returnable.
View return policy
Description
APRIL/TNFSF13 Polyclonal specifically detects APRIL/TNFSF13 in Human samples. It is validated for Western Blot.Specifications
| APRIL/TNFSF13 | |
| Polyclonal | |
| Biotin | |
| A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TNFSF13, TRDL1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8741 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| TNFSF13 | |
| E. coli-derived recombinant human APRIL aa 110-250 | |
| 50 μg | |
| Primary | |
| Detects human APRIL in Western blots. In this format, less than 1% cross-reactivity is observed with rhTRANCE, rhTNF-alpha , rhFas Ligand, rhTRAIL, rhGITR, and rhVEGI. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction