Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human APRIL/TNFSF13 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP182419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182419 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP182419 Supplier Novus Biologicals™ Supplier No. NBP182419
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human APRIL/TNFSF13 ELISA Kit (Colorimetric) measures human APRIL/TNFSF13 in cell culture supernatant, serum, plasma (edta, citrate, heparin).

Specifications

Assay Range 0.78 - 50.0ng/mL
Assay Sensitivity 0.4ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 8741
Gene Symbol TNFSF13
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human TNFSF13, Biotin-Conjugate anti-human TNFSF13 polyclonal antibody, Streptavidin-HRP, Human TNFSF13 Standard lyophilized, 100ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligan
Target APRIL/TNFSF13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.