Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ARA54 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5124
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5124 Supplier R&D Systems Supplier No. AF5124
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ARA54 Polyclonal specifically detects ARA54 in Human samples. It is validated for Western Blot.

Specifications

Antigen ARA54
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9UBS8
Gene Alias Androgen receptor-associated protein 54, ARA54androgen receptor associated protein 54, EC 6.3.2.-, HFB30, HFB30E3 ubiquitin-protein ligase RNF14, ring finger protein 14FLJ26004, RNF14, TRIAD2, Triad2 protein
Gene Symbols RNF14
Host Species Sheep
Immunogen E. coli-derived recombinant human ARA54 Met127-Gln261 Accession # Q9UBS8
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9604
Test Specificity Detects endogenous human ARA54 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.