Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Arginase 1/ARG1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC8026N
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Arginase 1/ARG1/liver Arginase | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| AI, ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver Arginase, Liver-type arginase, PGIF, Type I Arginase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 383 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 658922 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| P05089 | |
| ARG1 | |
| E. coli-derived recombinant human Arginase 1/ARG1 Met1-Lys322 Accession # P05089 | |
| 100 Tests | |
| Primary | |
| Detects human Arginase 1/ARG1 in ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction