Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Arginase 1/ARG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB58681
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Flow Cytometry, CyTOF-reported.Specifications
Arginase 1/ARG1/liver Arginase | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-reported | |
P05089 | |
ARG1 | |
E. coli-derived recombinant human Arginase 1/ARG1 Met1-Lys322 Accession # P05089 | |
100 μg | |
Primary | |
Detects human Arginase 1/ARG1 in ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
658922 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AI, ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver Arginase, Liver-type arginase, PGIF, Type I Arginase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
383 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction