Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ARNT/HIF-1 beta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5630
This item is not returnable.
View return policy
Description
ARNT/HIF-1 beta Polyclonal specifically detects ARNT/HIF-1 beta in Human samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.Specifications
ARNT/HIF-1 beta | |
Polyclonal | |
Unconjugated | |
P27540 | |
ARNT | |
E. coli-derived recombinant human ARNT/HIF-1 beta Asp517-Ala691 Accession # P27540 | |
100 μg | |
Primary | |
Detects human ARNT/HIF-1 beta in Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF1 beta, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO | |
Goat | |
Affinity Purified | |
RUO | |
405 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction