Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Arylsulfatase B/ARSB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4415
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4415 Supplier R&D Systems Supplier No. MAB4415
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Arylsulfatase B/ARSB Monoclonal specifically detects Arylsulfatase B/ARSB in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Arylsulfatase B/ARSB
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 708401
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15848
Gene Alias ARSB, arylsulfatase B, ASB, EC 3.1.6.12, G4S, MPS6, N-acetylgalactosamine-4-sulfatase
Gene Symbols ARSB
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Arylsulfatase B/ARSB Ser37-Met533 Accession # P15848
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 411
Test Specificity Detects human Arylsulfatase B/ARSB in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ARSA, rhARSJ, or recombinant mouse ARSG is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.