Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | ASAH2/N-acylsphingosine Amidohydrolase-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ASAH2/N-acylsphingosine Amidohydrolase-2 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, Nacylsphingosine Amidohydrolase2, NCDase, N-CDase | |
ASAH2 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
AAF86240 | |
56624 | |
Mouse myeloma cell line NS0-derived recombinant human ASAH2/N-acylsphingosine Amidohydrolase-2 Thr15-Ile761 Accession # AAF86240 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ASAH2/N-acylsphingosine Amidohydrolase-2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title