Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase |
---|---|
Clone | 511702 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Monoclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q02083 | |
27163 | |
Chinese hamster ovary cell line CHO-derived recombinant human ASAHL/N-acylethanolamine-hydrolyzing Acid A isoform 1 Ser29-Lys359 (Val151Ile) Accession # Q02083 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ASAHL/N-acylethanolamine-hydrolyzing Acid A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse ASAHL is observed and no cross-reactivity with recombinant human ASAH2 or recombinant mouse ASAH2 is observed. |
511702 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, NAAA, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, Nacylethanolaminehydrolyzing Acid Amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT | |
NAAA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title