Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ASGR1/ASGPR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$222.00 - $570.50
Specifications
Antigen | ASGPR1 |
---|---|
Clone | 950216 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4394SP
![]() |
R&D Systems
MAB4394SP |
25 μg |
Each for $222.00
|
|
|||||
MAB4394100
![]() |
R&D Systems
MAB4394100 |
100 μg |
Each for $570.50
|
|
|||||
Description
ASGPR1 Monoclonal specifically detects ASGPR1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
ASGPR1 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
P07306 | |
432 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
950216 | |
Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ASGPR, ASGPR 1, ASGP-R 1, ASGPR1, asialoglycoprotein receptor 1, CLEC4H1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1, MHL1, RHL1 | |
Mouse myeloma cell line NS0-derived recombinant human ASGR1/ASGPR1 Gln62-Leu291 Accession # P07306 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ASGR/1ASGPR1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title