Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ATAD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB9169100
This item is not returnable.
View return policy
Description
ATAD2 Monoclonal specifically detects ATAD2 in Human samples. It is validated for Immunocytochemistry.Specifications
| ATAD2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AAA nuclear coregulator cancer-associated protein, ANCCA, ANCCAMGC131938, ATPase family AAA domain-containing protein 2, ATPase family, AAA domain containing 2, CT137, DKFZp667N1320, EC 3.6.1.3, MGC142216, MGC29843, MGC5254, PRO2000 | |
| Human ATAD2 synthetic peptide Accession # Q6PL18 | |
| 100 μg | |
| Primary | |
| Detects human ATAD2 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 952947 | |
| Immunocytochemistry 8-25 ug/mL | |
| Q6PL18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 29028 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction