Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ATF6 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC71527V100UG
This item is not returnable.
View return policy
Description
ATF6 Monoclonal specifically detects ATF6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| ATF6 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ACHM7, activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6 alpha, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| E. coli-derived recombinant human ATF6, Met1-Thr192, Accession # P18850 | |
| 100 μg | |
| 22926 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2358C | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P18850 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human ATF6 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction