Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ATF6 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.16 - $520.35
Specifications
| Antigen | ATF6 |
|---|---|
| Clone | 2358J |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB715271SP
![]() |
R&D Systems
MAB715271SP |
25 μg |
Each for $200.16
|
|
|||||
MAB71527110
![]() |
R&D Systems
MAB715271100 |
100 μg |
Each for $520.35
|
|
|||||
Description
ATF6 Monoclonal antibody specifically detects ATF6 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| ATF6 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Rabbit | |
| Human | |
| P18850 | |
| 22926 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2358J | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACHM7, activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6 alpha, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| E. coli-derived recombinant human ATF6, Met1-Thr192, Accession # P18850 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title