Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ATG10 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5464SP
This item is not returnable.
View return policy
Description
ATG10 Polyclonal specifically detects ATG10 in Human samples. It is validated for Western Blot.Specifications
ATG10 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
Sheep | |
Affinity Purified | |
RUO | |
83734 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9H0Y0 | |
ATG10 | |
E. coli-derived recombinant human ATG10 Met1-Thr190 Accession # Q9H0Y0 | |
25 μg | |
Primary | |
Detects endogenous human ATG10 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction