Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATOX1 (NP_004036.1) Full-length Recombinant Protein expressed in yeast

Catalog No. 89935984 Shop All Abnova Corporation Products
Click to view available options
Quantity:
100 μg

Used for SDS-PAGE

This gene encodes a copper chaperone that plays a role in copper homeostasis by binding and transporting cytosolic copper to ATPase proteins in the trans-Golgi network for later incorporation to the ceruloplasmin. This protein also functions as an antioxidant against superoxide and hydrogen peroxide, and therefore, may play a significant role in cancer carcinogenesis. Because of its cytogenetic location, this gene represents a candidate gene for 5q-syndrome. [provided by RefSeq]

Sequence: MPKHEFSVDMTCGGCAEAVSRVLNKLGGVKYDIDLPNKKVCIESEHSMDTLLATLKKTGKTVSYLGLE

Specifications

Accession Number NP_004036.1
For Use With (Application) SDS-PAGE
Formulation In 50mM HEPES, 100mM NaCl, pH 7.5 (30% glycerol, 30mM Glutathione, 0.5% TritonX-100, 1mM dithiothreitol)
Gene ID (Entrez) 475
Molecular Weight (g/mol) 7.39kDa
Name ATOX1 (Human) Recombinant Protein
Preparation Method Yeast expression system
Purification Method Affinity Purification
Quantity 100 μg
Storage Requirements Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATX1/HAH1/MGC138453/MGC138455
Common Name ATOX1
Gene Symbol ATOX1
Species Yeast
Recombinant Recombinant
Protein Tag None
Expression System Yeast expression system
Form Liquid
Purity or Quality Grade 0.9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.