Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Attractin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7238
This item is not returnable.
View return policy
Description
Attractin Polyclonal specifically detects Attractin in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Attractin | |
Polyclonal | |
Unconjugated | |
NP_647538 | |
ATRN | |
Chinese hamster ovary cell line CHO-derived recombinant human Attractin Ala84-Gln1272 Accession # NP_647538 | |
100 μg | |
Primary | |
Detects human Attractin in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
ATRN, attractin, DPPT-L, KIAA0548, Mahogany Protein, MGC126754, MGCA | |
Sheep | |
Affinity Purified | |
RUO | |
8455 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction