Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Autoimmune Regulator/AIRE Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5936
This item is not returnable.
View return policy
Description
Autoimmune Regulator/AIRE Polyclonal specifically detects Autoimmune Regulator/AIRE in Human samples. It is validated for Western Blot.Specifications
Autoimmune Regulator/AIRE | |
Polyclonal | |
Unconjugated | |
O43918 | |
AIRE | |
E. coli-derived recombinant human AIRE Ser476-Ser545 Accession # O43918 | |
100 μg | |
Primary | |
Detects human AIRE in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AIRE, APECED, APECED protein, APECEDAPSI, APS1, APSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 | |
Goat | |
Affinity Purified | |
RUO | |
326 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction