Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Axl Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB154G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB154G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB154G Supplier R&D Systems Supplier No. FAB154G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Axl Monoclonal specifically detects Axl in Human samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen Axl
Applications Flow Cytometry, KnockDown
Classification Monoclonal
Clone 108724
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells, Knockout Validated 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAA61243
Gene Alias AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL
Gene Symbols AXL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Axl Met1-Pro440 Accession # AAA61243
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 558
Test Specificity Detects human Axl in direct ELISAs. In direct ELISAs, this antibody shows no cross-reactivity with recombinant mouse Axl, recombinant human (rh) Dtk or rhMer.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.