Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Axl Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB154RR100UG
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Human samples. It is validated for Flow Cytometry.Specifications
Axl | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
Mouse myeloma cell line NS0-derived recombinant human Axl Met1-Pro440 Accession # AAA61243 | |
100 μg | |
558 | |
Human | |
Supernatant |
Flow Cytometry | |
108724R | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAA61243 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Axl in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction