Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B-Raf Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3424SP
This item is not returnable.
View return policy
Description
B-Raf Polyclonal specifically detects B-Raf in Human samples. It is validated for Western Blot, Simple Western.Specifications
B-Raf | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
94 kDa B-raf protein, BRaf, B-Raf proto-oncogene serine/threonine-protein kinase (p94), BRAF1, B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
Goat | |
Affinity Purified | |
RUO | |
673 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
P15056 | |
BRAF | |
E. coli-derived recombinant human B-Raf Met1-His766 Accession # P15056 | |
25 μg | |
Primary | |
Detects human B-Raf in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction