Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-1/CD80 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF140
This item is not returnable.
View return policy
Description
B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
B7-1/CD80 | |
Polyclonal | |
Unconjugated | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Goat | |
Affinity Purified | |
RUO | |
941 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CD80 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-1/CD80 Extracellular domain | |
100 μg | |
Primary | |
Detects human B7-1/CD80 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human B7-2 and recombinant mouse B7-2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction