Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-2/CD86 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB141U100UG
This item is not returnable.
View return policy
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human samples. It is validated for Flow Cytometry.Specifications
B7-2/CD86 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P42081 | |
CD86 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86 Ala23-His244 Accession # P42081 | |
RUO | |
942 | |
Human | |
IgG1 |
Flow Cytometry | |
37301 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
Mouse | |
100 μg | |
Primary | |
Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, recombinant mouse B7-2, recombinant rat B7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b, rhB7-H4, or rhB7-L2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction