Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-2/CD86 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
| Antigen | B7-2/CD86 |
|---|---|
| Clone | 1036411 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB108522SP
![]() |
R&D Systems
MAB108522SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10852210
![]() |
R&D Systems
MAB108522100 |
100 μg |
Each for $432.50
|
|
|||||
Description
B7-2/CD86 Monoclonal antibody specifically detects B7-2/CD86 in Human samples. It is validated for nanSpecifications
| B7-2/CD86 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Human embryonic kidney cell, HEK293 derived human B7-2/CD86, Leu26-Pro247, Accession # NP_787058 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1036411 | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_787058 | |
| 942 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title