Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF165SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF165SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF165SP Supplier R&D Systems Supplier No. AF165SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

B7-H2/ICOSLG Polyclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen B7-H2/ICOSLG
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75144
Gene Alias B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Gene Symbols ICOSLG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H2 Asp19-Ser258 Accession # O75144
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23308
Test Specificity Detects human B7-H2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse B7-H2 and recombinant human B7-H1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.