Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB165P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB165P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB165P Supplier R&D Systems Supplier No. FAB165P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H2/ICOSLG
Applications Flow Cytometry
Classification Monoclonal
Clone 136726
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O75144
Gene Alias B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand
Gene Symbols ICOSLG
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H2 Asp19-Ser258 Accession # O75144
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23308
Test Specificity Detects human B7-H2 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) B7-1, rhB7-2, rhB7-H1, rhB7-H3, or recombinant mouse B7-H2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.