Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H3 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1027V025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1027V025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1027V025 Supplier R&D Systems Supplier No. FAB1027V025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H3/CD276
Applications Flow Cytometry
Classification Monoclonal
Clone 185504
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_079516
Gene Alias B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule
Gene Symbols CD276
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H3 Leu29-Pro245 Accession # NP_079516
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80381
Test Specificity Detects human B7-H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) B7-H1, rhB7-H2, rhB7-1, rhB7-2, or recombinant mouse PD-L2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.