Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H3 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1027G100UG
This item is not returnable.
View return policy
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Flow Cytometry.Specifications
B7-H3/CD276 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_079516 | |
CD276 | |
Mouse myeloma cell line NS0-derived recombinant human B7-H3 Leu29-Pro245 Accession # NP_079516 | |
RUO | |
80381 | |
Human | |
IgG1 |
Flow Cytometry | |
185504 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Mouse | |
100 μg | |
Primary | |
Detects human B7-H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) B7-H1, rhB7-H2, rhB7-1, rhB7-2, or recombinant mouse PD-L2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction