Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Supplier: R&D Systems AF1027SP
This item is not returnable.
View return policy
Description
B7-H3/CD276 Polyclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, CyTOF-ready, Knockout Validated.Specifications
B7-H3/CD276 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Goat | |
Affinity Purified | |
RUO | |
80381 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, KnockDown | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, Knockout Validated | |
NP_079516 | |
CD276 | |
Mouse myeloma cell line NS0-derived recombinant human B7-H3 Leu29-Pro245 Accession # NP_079516 | |
25 μg | |
Primary | |
Detects human B7-H3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) B7-H2 is observed, and less than 1% cross-reactivity with rhB7-1, rhB7-2, rhB7-H1, and rhPD-L2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction