Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H3 MAb (Clone 185504), R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$241.00 - $2070.00
Specifications
Antigen | B7-H3/CD276 |
---|---|
Clone | 185504 |
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated |
Applications | Western Blot, Flow Cytometry, CyTOF, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1027SP
![]() |
R&D Systems
MAB1027SP |
25 μg |
Each for $241.00
|
|
|||||
MAB1027100
![]() |
R&D Systems
MAB1027100 |
100 μg |
Each for $584.50
|
|
|||||
MAB1027500
![]() |
R&D Systems
MAB1027500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated.Specifications
B7-H3/CD276 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF, KnockDown | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
CD276 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
185504 | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated | |
Monoclonal | |
Mouse | |
Human | |
NP_079516 | |
80381 | |
Mouse myeloma cell line NS0-derived recombinant human B7-H3 Leu29-Pro245 Accession # NP_079516 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human B7-H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) B7-H1, rhB7-H2, rhB7-1, rhB7-2, or recombinant mouse PD-L2 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title