Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6576U100UG
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal specifically detects B7-H4 in Human samples. It is validated for Flow Cytometry.Specifications
B7-H4 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
NS0 mouse myeloma cell line transfected with human B7-H4 Phe29-Ser258 Accession # Q727D3 | |
RUO | |
79679 | |
Human | |
IgG2b |
Flow Cytometry | |
935317 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q727D3 | |
Mouse | |
100 μg | |
Primary | |
Stains human B7-H4 transfectants but not irrelevant transfectants in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction