Learn More
Human B7-H4 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | B7-H4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2318A |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q7Z7D3 |
| Gene Alias | B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.