Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $538.00
Specifications
Antigen | B7-H4 |
---|---|
Clone | 973815 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB65763SP
![]() |
R&D Systems
MAB65763SP |
25 μg |
Each for $200.50
|
|
|||||
MAB65763100
![]() |
R&D Systems
MAB65763100 |
100 μg |
Each for $538.00
|
|
|||||
Description
B7-H4 Monoclonal specifically detects B7-H4 in Human samples. It is validated for ELISA.Specifications
B7-H4 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q7Z7D3 | |
79679 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
973815 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
Mouse myeloma cell line NS0-derived recombinant human B7-H4 Phe29-Ala258 Accession # Q7Z7D3 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human B7-H4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title