Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6576SP
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal specifically detects B7-H4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
B7-H4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79679 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
935317 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q727D3 | |
VTCN1 | |
NS0 mouse myeloma cell line transfected with human B7-H4 Phe29-Ser258 Accession # Q727D3 | |
25 μg | |
Primary | |
Stains human B7-H4 transfectants but not irrelevant transfectants in flow cytometry. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction