Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H6 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB71442S10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB71442S10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB71442S10 Supplier R&D Systems Supplier No. FAB71442S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen B7-H6
Applications Flow Cytometry
Classification Monoclonal
Clone 875002
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q68D85
Gene Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H6 Met1-Ser262 Accession # Q68D85
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 374383
Test Specificity Detects human B7-H6 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.