Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7144
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7144 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7144 Supplier R&D Systems Supplier No. MAB7144
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated.

Order Info

The product is shipped at ambient temperature.

Specifications

Antigen B7-H6
Applications Flow Cytometry, ELISA, CyTOF, Immunoassay
Classification Monoclonal
Clone 875001
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q68D85
Gene Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Gene Symbols NCR3LG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H6 Met1-Ser262 Accession # Q68D85
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374383
Test Specificity Detects human B7-H6 in ELISA.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.