Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB71444SP
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Blocking AssaySpecifications
B7-H6 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1 | |
Human embryonic kidney cell HEK293-derived human B7-H6, Asp25-Ser262, Accession # Q68D85 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Blocking Assay | |
1018457 | |
Blockade of Receptor-ligand Interaction | |
Q68D85 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
374383 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction