Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H6 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7144P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7144P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7144P100 Supplier R&D Systems Supplier No. FAB7144P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry, Knockout Validated.

Specifications

Antigen B7-H6
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 875001
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q68D85
Gene Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Gene Symbols NCR3LG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H6 Met1-Ser262 Accession # Q68D85
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374383
Test Specificity Detects human B7-H6 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.