Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human B7-H7/HHLA2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB80841U100UG
This item is not returnable.
View return policy
Description
B7-H7/HHLA2 Monoclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry.Specifications
B7-H7/HHLA2 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44 | |
100 μg | |
11148 | |
Human | |
Supernatant |
Flow Cytometry | |
907812 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9UM44 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human B7-H7/HHLA2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction