Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B7-H7/HHLA2 Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody

$200.50 - $549.50

Specifications

Antigen B7-H7/HHLA2
Dilution Western Blot 1 ug/mL, Immunocytochemistry 1-15 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AF8084SP
GREENER_CHOICE
View Documents
R&D Systems
AF8084SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
AF8084100
GREENER_CHOICE
View Documents
R&D Systems
AF8084100
100 μg
Each for $549.50
Only null left
Add to Cart
 
Description

Description

B7-H7/HHLA2 Polyclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

B7-H7/HHLA2
Western Blot, Immunocytochemistry
Unconjugated
Goat
Human
Q9UM44
11148
IgG
Antigen Affinity-purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot 1 ug/mL, Immunocytochemistry 1-15 ug/mL
Polyclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2 Met1-Asn344 Accession # Q9UM44
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects human B7-H7/HHLA2 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.