Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bad Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6405
This item is not returnable.
View return policy
Description
Bad Monoclonal specifically detects Bad in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.Specifications
Bad | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 5-25 ug/mL, Knockout Validated | |
Q92934 | |
BAD | |
E. coli-derived recombinant human Bad Met1-Gln168 Accession # Q92934 | |
100 μg | |
Primary | |
Detects human Bad in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Bad (aa 1-130), rmBcl-2 alpha , recombinant human (rh) Bcl-2, or rhBCL2L12 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
612312 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
572 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction