Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Description
The Human BAFF/BLyS/TNFSF13B Quantikine ELISA Kit quantifies human BAFF/BLyS/TNFSF13B in cell culture supernates (25 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
Specifications
Specifications
Assay Range | 62.50 - 4000pg/mL |
Assay Sensitivity | 6.44pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
Specificity | Natural and Recombinant Human BAFF/BLyS |
Sufficient For | 1 x 96-well Plate |
Target Species | Human |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction