Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BAFFR/TNFRSF13C Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1162V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1162V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1162V100 Supplier R&D Systems Supplier No. FAB1162V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry
Classification Monoclonal
Clone 2403C
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Gene Accession No. Q96RJ3
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human BAFF R/TNFRSF13C, Ser7-Ala71, Accession No. Q96RJ3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 115650
Test Specificity Detects human BAFF R/TNFRSF13C in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.