Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BAFFR/TNFRSF13C APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1162A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1162A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1162A Supplier R&D Systems Supplier No. FAB1162A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BAFFR/TNFRSF13C Polyclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q96RJ3
Gene Symbols TNFRSF13C
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human BAFF R/TNFRSF13C Arg2-Ala71 Accession # Q96RJ3
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115650
Test Specificity Detects human BAFF R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse BAFF R and less than 2% cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, rhRANK, rhTNF RI, and rhTNF RII is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.