Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAI1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB49691V100UG
This item is not returnable.
View return policy
Description
BAI1 Monoclonal specifically detects BAI1 in Human samples. It is validated for Flow Cytometry.Specifications
BAI1 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
HEK293 human embryonic kidney cell line transfected with human BAI1, Ala31-Thr879, Accession # O14514 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
480912 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O14514 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
575 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction