Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAI2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | BAI2 |
---|---|
Clone | 2528B |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9338SP
![]() |
R&D Systems
MAB9338SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9338100
![]() |
R&D Systems
MAB9338100 |
100 μg |
Each for $467.50
|
|
|||||
Description
BAI2 Monoclonal specifically detects BAI2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
BAI2 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2 | |
Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2528B | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
O60241-3 | |
576 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title