Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAI3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3965G100UG
This item is not returnable.
View return policy
Description
BAI3 Monoclonal specifically detects BAI3 in Human samples. It is validated for Flow Cytometry.Specifications
BAI3 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242 | |
100 μg | |
577 | |
Human | |
Supernatant |
Flow Cytometry | |
409611 | |
Flow Cytometry 0.25-1 μg/106 cells | |
O60242 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human BAI3 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction