Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAI3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB39651
This item is not returnable.
View return policy
Description
BAI3 Monoclonal specifically detects BAI3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
BAI3 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
O60242 | |
BAI3 | |
Mouse myeloma cell line NS0-derived recombinant human BAI3 Ala25-Thr880 Accession # O60242 | |
100 μg | |
Primary | |
Detects human BAI3 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
409633 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
577 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction