Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAK Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF816
This item is not returnable.
View return policy
Description
BAK Polyclonal specifically detects BAK in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
BAK | |
Polyclonal | |
Unconjugated | |
Q16611 | |
BAK1 | |
KLH-coupled human BAK synthetic peptide AAPADPEMVTLPLQPSSTMGC Accession # Q16611 | |
100 μg | |
Primary | |
Detects human BAK in Western blots. This antibody does not detect mouse BAK. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1-4 ug/mL | |
Apoptosis regulator BAK, BAK1, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
Rabbit | |
Affinity Purified | |
RUO | |
578 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction